Assay Details
| Gene Symbol: LOC4338448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783697.2 | XP_015639183.1 | 9 - 10 | 1054 | 100 | |
| XM_015783698.2 | XP_015639184.1 | 9 - 10 | 1073 | 100 | ||
| GenBank mRNA | D38232.1 | - | 8 - 9 | 697 | 100 | |
More Information
Set Membership: |
|

LOC4338448
Genomic Map