Assay Details
| Gene Symbol: LOC4338566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782276.2 | XP_015637762.1 | 3 - 4 | 459 | 73 | |
| GenBank mRNA | AK108439.1 | - | 3 - 4 | 440 | 73 | |
More Information
Set Membership: |
|

LOC4338566
Genomic Map