Assay Details
| Gene Symbol: LOC4338646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785117.1 | XP_015640603.1 | 3 - 4 | 1040 | 56 | |
| GenBank mRNA | AK061728.1 | - | 3 - 4 | 1045 | 56 | |
| AK065038.1 | - | 3 - 4 | 849 | 56 | ||
More Information
Set Membership: |
|

LOC4338646
Genomic Map