Assay Details
| Gene Symbol: LOC4338659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK071425.1 | - | 3 - 4 | 881 | 112 | |
| AK073634.1 | - | 3 - 4 | 898 | 112 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338659| Gene Symbol: LOC4338659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK071425.1 | - | 3 - 4 | 881 | 112 | |
| AK073634.1 | - | 3 - 4 | 898 | 112 | ||
Set Membership: |
|