Assay Details
| Gene Symbol: LOC4338768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785019.2 | XP_015640505.1 | 1 - 2 | 379 | 140 | |
| GenBank mRNA | AK058950.1 | - | 1 - 2 | 292 | 140 | |
| AK059821.1 | - | 1 - 2 | 364 | 140 | ||
| AK064944.1 | - | 1 - 2 | 287 | 140 | ||
| AK104851.1 | - | 1 - 2 | 246 | 140 | ||
| AK106750.1 | - | 1 - 2 | 271 | 140 | ||
More Information
Set Membership: |
|

LOC4338768
Genomic Map