Assay Details
| Gene Symbol: LOC4338768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785019.2 | XP_015640505.1 | 5 - 6 | 1376 | 89 | |
| GenBank mRNA | AK064944.1 | - | 5 - 6 | 1284 | 89 | |
| AK104851.1 | - | 5 - 6 | 1243 | 89 | ||
More Information
Set Membership: |
|

LOC4338768
Genomic Map