Assay Details
| Gene Symbol: LOC4338768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785019.2 | XP_015640505.1 | 8 - 9 | 2253 | 94 | |
| GenBank mRNA | AK064944.1 | - | 8 - 9 | 2161 | 94 | |
| AK104851.1 | - | 8 - 9 | 2120 | 94 | ||
More Information
Set Membership: |
|

LOC4338768
Genomic Map