Assay Details
| Gene Symbol: LOC4338779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784701.2 | XP_015640187.1 | 2 - 3 | 1078 | 61 | |
| GenBank mRNA | AK070558.1 | - | 2 - 3 | 994 | 61 | |
| AK119621.1 | - | 2 - 3 | 993 | 61 | ||
More Information
Set Membership: |
|

LOC4338779
Genomic Map