Assay Details
| Gene Symbol: LOC4338850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK066897.1 | - | 3 - 4 | 910 | 119 | |
| AK101519.1 | - | 3 - 4 | 978 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338850| Gene Symbol: LOC4338850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK066897.1 | - | 3 - 4 | 910 | 119 | |
| AK101519.1 | - | 3 - 4 | 978 | 119 | ||
Set Membership: |
|