Assay Details
| Gene Symbol: LOC4338901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784530.2 | XP_015640016.1 | 11 - 12 | 1201 | 81 | |
| GenBank mRNA | AB488786.1 | - | 9 - 10 | 866 | 81 | |
| AK060776.1 | - | 7 - 8 | 695 | 81 | ||
| AK120229.1 | - | 11 - 12 | 1579 | 81 | ||
More Information
Set Membership: |
|

LOC4338901
Genomic Map