Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 14 - 15 | 1934 | 68 | |
| GenBank mRNA | AK102633.1 | - | 14 - 15 | 1828 | 68 | |
| AY574031.1 | - | 14 - 15 | 1710 | 68 | ||
| D49714.1 | - | 14 - 15 | 1808 | 68 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map