Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 15 - 16 | 2095 | 123 | |
| GenBank mRNA | AK102633.1 | - | 15 - 16 | 1989 | 123 | |
| AY574031.1 | - | 15 - 16 | 1871 | 123 | ||
| D49714.1 | - | 15 - 16 | 1969 | 123 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map