Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 16 - 17 | 2156 | 94 | |
| GenBank mRNA | AK102633.1 | - | 16 - 17 | 2050 | 94 | |
| AY574031.1 | - | 16 - 17 | 1932 | 94 | ||
| D49714.1 | - | 16 - 17 | 2030 | 94 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map