Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 19 - 20 | 2378 | 70 | |
| GenBank mRNA | AK102633.1 | - | 19 - 20 | 2272 | 70 | |
| D49714.1 | - | 19 - 20 | 2252 | 70 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map