Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 3 - 4 | 563 | 72 | |
| GenBank mRNA | AK101985.1 | - | 3 - 4 | 457 | 72 | |
| AK102633.1 | - | 3 - 4 | 457 | 72 | ||
| AY574031.1 | - | 3 - 4 | 339 | 72 | ||
| D49714.1 | - | 3 - 4 | 437 | 72 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map