Assay Details
| Gene Symbol: LOC4338979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784690.2 | XP_015640176.1 | 6 - 7 | 1150 | 89 | |
| GenBank mRNA | AK101985.1 | - | 6 - 7 | 1044 | 89 | |
| AK102633.1 | - | 6 - 7 | 1044 | 89 | ||
| AY574031.1 | - | 6 - 7 | 926 | 89 | ||
| D49714.1 | - | 6 - 7 | 1024 | 89 | ||
More Information
Set Membership: |
|

LOC4338979
Genomic Map