Assay Details
Gene Symbol: LOC4339268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784027.2 | XP_015639513.1 | 10 - 11 | 1609 | 130 | |
GenBank mRNA | AK101520.1 | - | 10 - 11 | 1567 | 130 | |
AK105718.1 | - | 3 - 4 | 352 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
