Assay Details
| Gene Symbol: LOC4339268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784027.2 | XP_015639513.1 | 4 - 5 | 621 | 97 | |
| GenBank mRNA | AK101520.1 | - | 4 - 5 | 579 | 97 | |
More Information
Set Membership: |
|

LOC4339268
Genomic Map