Assay Details
| Gene Symbol: LOC4339274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782337.2 | XP_015637823.1 | 6 - 7 | 1095 | 100 | |
| GenBank mRNA | AB115419.1 | - | 6 - 7 | 998 | 100 | |
| AK068958.1 | - | 6 - 7 | 1096 | 100 | ||
More Information
Set Membership: |
|

LOC4339274
Genomic Map