Assay Details
| Gene Symbol: LOC4339446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783773.2 | XP_015639259.1 | 2 - 3 | 224 | 62 | |
| GenBank mRNA | AJ251562.1 | - | 2 - 3 | 165 | 62 | |
| AK071833.1 | - | 2 - 3 | 209 | 62 | ||
More Information
Set Membership: |
|

LOC4339446
Genomic Map