Assay Details
| Gene Symbol: LOC4339446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783773.2 | XP_015639259.1 | 5 - 6 | 640 | 84 | |
| GenBank mRNA | AJ251562.1 | - | 5 - 6 | 581 | 84 | |
| AK071833.1 | - | 5 - 6 | 625 | 84 | ||
More Information
Set Membership: |
|

LOC4339446
Genomic Map