Assay Details
| Gene Symbol: LOC4339460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783723.2 | XP_015639209.1 | 4 - 5 | 507 | 96 | |
| GenBank mRNA | AK071120.1 | - | 4 - 5 | 447 | 96 | |
| AK105671.1 | - | 4 - 5 | 438 | 96 | ||
More Information
Set Membership: |
|

LOC4339460
Genomic Map