Assay Details
| Gene Symbol: LOC4339675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784733.2 | XP_015640219.1 | 12 - 13 | 1093 | 83 | |
| GenBank mRNA | AK070411.1 | - | 12 - 13 | 1092 | 83 | |
More Information
Set Membership: |
|

LOC4339675
Genomic Map