Assay Details
| Gene Symbol: LOC4339675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784733.2 | XP_015640219.1 | 13 - 14 | 1153 | 64 | |
| GenBank mRNA | AK070411.1 | - | 13 - 14 | 1152 | 64 | |
More Information
Set Membership: |
|

LOC4339675
Genomic Map