Assay Details
| Gene Symbol: LOC4339675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784733.2 | XP_015640219.1 | 8 - 9 | 849 | 97 | |
| GenBank mRNA | AK070411.1 | - | 8 - 9 | 848 | 97 | |
More Information
Set Membership: |
|

LOC4339675
Genomic Map