Assay Details
| Gene Symbol: LOC4339765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781969.2 | XP_015637455.1 | 6 - 7 | 2248 | 62 | |
| GenBank mRNA | AK120811.1 | - | 5 - 6 | 2291 | 62 | |
More Information
Set Membership: |
|

LOC4339765
Genomic Map