Assay Details
| Gene Symbol: LOC4339802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783742.1 | XP_015639228.1 | 21 - 22 | 4431 | 75 | |
| GenBank mRNA | AK106591.1 | - | 6 - 7 | 807 | 75 | |
More Information
Set Membership: |
|

LOC4339802
Genomic Map