Assay Details
| Gene Symbol: LOC4339910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787279.2 | XP_015642765.1 | 10 - 11 | 1467 | 68 | |
| XM_015787280.2 | XP_015642766.1 | 10 - 11 | 1456 | 68 | ||
| GenBank mRNA | AK060117.1 | - | 4 - 5 | 456 | 68 | |
| AK062098.1 | - | 8 - 9 | 1130 | 68 | ||
| AK101334.1 | - | 10 - 11 | 1462 | 68 | ||
| EU325985.1 | - | 9 - 10 | 1373 | 68 | ||
More Information
Set Membership: |
|

LOC4339910
Genomic Map