Assay Details
| Gene Symbol: LOC4339910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787279.2 | XP_015642765.1 | 6 - 7 | 918 | 79 | |
| XM_015787280.2 | XP_015642766.1 | 6 - 7 | 907 | 79 | ||
| GenBank mRNA | AK062098.1 | - | 4 - 5 | 581 | 79 | |
| EU325985.1 | - | 5 - 6 | 824 | 79 | ||
More Information
Set Membership: |
|

LOC4339910
Genomic Map