Assay Details
| Gene Symbol: LOC4339910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787279.2 | XP_015642765.1 | 8 - 9 | 1326 | 107 | |
| XM_015787280.2 | XP_015642766.1 | 8 - 9 | 1315 | 107 | ||
| GenBank mRNA | AK060117.1 | - | 2 - 3 | 315 | 107 | |
| AK062098.1 | - | 6 - 7 | 989 | 107 | ||
| AK101334.1 | - | 8 - 9 | 1321 | 107 | ||
| EU325985.1 | - | 7 - 8 | 1232 | 107 | ||
More Information
Set Membership: |
|

LOC4339910
Genomic Map