Assay Details
| Gene Symbol: LOC4340087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787003.2 | XP_015642489.1 | 2 - 3 | 1366 | 118 | |
| GenBank mRNA | AK242105.1 | - | 1 - 2 | 353 | 118 | |
More Information
Set Membership: |
|

LOC4340087
Genomic Map