Assay Details
| Gene Symbol: LOC4340223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787481.2 | XP_015642967.1 | 4 - 5 | 1044 | 98 | |
| GenBank mRNA | AK073970.1 | - | 4 - 5 | 1012 | 98 | |
More Information
Set Membership: |
|

LOC4340223
Genomic Map