Assay Details
| Gene Symbol: LOC4340567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788760.2 | XP_015644246.1 | 3 - 4 | 1258 | 80 | |
| GenBank mRNA | AB115915.1 | - | 3 - 4 | 1074 | 80 | |
| AB115916.1 | - | 3 - 4 | 1060 | 80 | ||
| AK101978.1 | - | 3 - 4 | 1224 | 80 | ||
More Information
Set Membership: |
|

LOC4340567
Genomic Map