Assay Details
| Gene Symbol: LOC4340567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788760.2 | XP_015644246.1 | 7 - 8 | 1673 | 85 | |
| GenBank mRNA | AB115915.1 | - | 7 - 8 | 1489 | 85 | |
| AB115916.1 | - | 7 - 8 | 1475 | 85 | ||
| AK101978.1 | - | 7 - 8 | 1639 | 85 | ||
More Information
Set Membership: |
|

LOC4340567
Genomic Map