Assay Details
| Gene Symbol: LOC4340590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785837.2 | XP_015641323.1 | 2 - 3 | 596 | 63 | |
| GenBank mRNA | AK061102.1 | - | 2 - 3 | 339 | 63 | |
| AK063083.1 | - | 2 - 3 | 380 | 63 | ||
| AK067253.1 | - | 2 - 3 | 372 | 63 | ||
More Information
Set Membership: |
|

LOC4340590
Genomic Map