Assay Details
| Gene Symbol: LOC4340590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785837.2 | XP_015641323.1 | 5 - 6 | 831 | 103 | |
| GenBank mRNA | AK061102.1 | - | 5 - 6 | 574 | 103 | |
More Information
Set Membership: |
|

LOC4340590
Genomic Map