Assay Details
| Gene Symbol: LOC4340590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785837.2 | XP_015641323.1 | 6 - 7 | 901 | 66 | |
| GenBank mRNA | AK061102.1 | - | 6 - 7 | 644 | 66 | |
| AK067253.1 | - | 4 - 5 | 2252 | 66 | ||
More Information
Set Membership: |
|

LOC4340590
Genomic Map