Assay Details
| Gene Symbol: LOC4341066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786515.2 | XP_015642001.1 | 3 - 4 | 1738 | 87 | |
| GenBank mRNA | AK100169.1 | - | 3 - 4 | 1730 | 87 | |
More Information
Set Membership: |
|

LOC4341066
Genomic Map