Assay Details
| Gene Symbol: LOC4341211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787162.2 | XP_015642648.1 | 9 - 10 | 1446 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4341211| Gene Symbol: LOC4341211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787162.2 | XP_015642648.1 | 9 - 10 | 1446 | 120 | |
Set Membership: |
|