Assay Details
| Gene Symbol: LOC4342077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787016.1 | XP_015642502.1 | 1 - 2 | 199 | 131 | |
| GenBank mRNA | AK063385.1 | - | 1 - 2 | 144 | 131 | |
| AK102478.1 | - | 1 - 2 | 148 | 131 | ||
| GU120345.1 | - | 1 - 2 | 125 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787016.1 transcripts.
Set Membership: |
|

LOC4342077
Genomic Map