Assay Details
| Gene Symbol: LOC4342077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787016.1 | XP_015642502.1 | 7 - 8 | 928 | 100 | |
| GenBank mRNA | AK062023.1 | - | 2 - 3 | 199 | 100 | |
| AK063385.1 | - | 7 - 8 | 873 | 100 | ||
| GU120345.1 | - | 7 - 8 | 854 | 100 | ||
More Information
Set Membership: |
|

LOC4342077
Genomic Map