Assay Details
| Gene Symbol: LOC4342103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788229.2 | XP_015643715.1 | 12 - 13 | 7237 | 87 | |
| GenBank mRNA | AK109634.1 | - | 1 - 2 | 456 | 87 | |
More Information
Set Membership: |
|

LOC4342103
Genomic Map