Assay Details
| Gene Symbol: LOC4342103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788229.2 | XP_015643715.1 | 14 - 15 | 8052 | 69 | |
| GenBank mRNA | AK109634.1 | - | 3 - 4 | 1271 | 69 | |
More Information
Set Membership: |
|

LOC4342103
Genomic Map