Assay Details
| Gene Symbol: LOC4342129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787785.2 | XP_015643271.1 | 6 - 7 | 524 | 72 | |
| XM_015787786.2 | XP_015643272.1 | 6 - 7 | 516 | 72 | ||
| GenBank mRNA | AF093632.1 | - | 6 - 7 | 419 | 72 | |
| AK062226.1 | - | 6 - 7 | 492 | 72 | ||
| AK068898.1 | - | 5 - 6 | 577 | 72 | ||
| BT129146.1 | - | 6 - 7 | 488 | 72 | ||
| BT129798.1 | - | 3 - 4 | 151 | 72 | ||
| BT130172.1 | - | 6 - 7 | 478 | 72 | ||
| BT131862.1 | - | 4 - 5 | 325 | 72 | ||
| EU847026.1 | - | 6 - 7 | 445 | 72 | ||
More Information
Set Membership: |
|

LOC4342129
Genomic Map