Assay Details
| Gene Symbol: LOC4342205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791217.2 | XP_015646703.1 | 1 | 3423 | 101 | |
| GenBank mRNA | AK064359.1 | - | 1 | 1931 | 101 | |
| AK121984.1 | - | 1 | 3246 | 101 | ||
| Gene Symbol: LOC4330264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK103166.1 | - | 1 | 2784 | 101 | |
More Information
Set Membership: |
|

LOC4342205
Genomic Map