Assay Details
| Gene Symbol: LOC4342300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792376.2 | XP_015647862.1 | 13 - 14 | 1521 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4342300| Gene Symbol: LOC4342300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792376.2 | XP_015647862.1 | 13 - 14 | 1521 | 105 | |
Set Membership: |
|