Assay Details
| Gene Symbol: LOC4343098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789128.2 | XP_015644614.1 | 2 - 3 | 779 | 82 | |
| GenBank mRNA | AK073296.1 | - | 2 - 3 | 474 | 82 | |
More Information
Set Membership: |
|

LOC4343098
Genomic Map