Assay Details
| Gene Symbol: LOC4343161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792121.2 | XP_015647607.1 | 3 - 4 | 1123 | 84 | |
| GenBank mRNA | AK101767.1 | - | 3 - 4 | 949 | 84 | |
More Information
Set Membership: |
|

LOC4343161
Genomic Map