Assay Details
| Gene Symbol: LOC4343161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792121.2 | XP_015647607.1 | 5 - 6 | 1598 | 114 | |
| GenBank mRNA | AK101767.1 | - | 5 - 6 | 1424 | 114 | |
| BT129831.1 | - | 1 - 2 | 55 | 114 | ||
More Information
Set Membership: |
|

LOC4343161
Genomic Map