Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 11 - 12 | 1806 | 83 | |
| XM_015791757.2 | XP_015647243.1 | 11 - 12 | 1808 | 83 | ||
| XM_015791758.2 | XP_015647244.1 | 11 - 12 | 1748 | 83 | ||
| XM_015791759.2 | XP_015647245.1 | 11 - 12 | 1808 | 83 | ||
| XM_015791760.2 | XP_015647246.1 | 11 - 12 | 1806 | 83 | ||
| XM_015791761.2 | XP_015647247.1 | 11 - 12 | 1804 | 83 | ||
| XM_015791762.2 | XP_015647248.1 | 11 - 12 | 1805 | 83 | ||
| XM_015791763.2 | XP_015647249.1 | 11 - 12 | 1760 | 83 | ||
| XM_015791764.2 | XP_015647250.1 | 11 - 12 | 1810 | 83 | ||
| XM_015791765.2 | XP_015647251.1 | 11 - 12 | 1810 | 83 | ||
| GenBank mRNA | AK067461.1 | - | 11 - 12 | 83 | ||
More Information
Set Membership: |
|

LOC4343276
Genomic Map